accessibility menu, dialog, popup

UserName

CAS RN 936-49-2

IUPAC Name: 2-phenyl-4,5-dihydro-1H-imidazol-3-ium
Synonyms: 2-phenyl-4,5-dihydro-3H-imidazol-1-ium
Molecular Weight (g/mol): 147.2
Molecular Formula: C9H11N2
InChi Key: BKCCAYLNRIRKDJ-UHFFFAOYSA-O
SMILES: C1C[NH+]=C(N1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)