CAS RN 94-08-6
CAS RN 94-08-6
IUPAC Name:
ethyl 4-methylbenzoate
Synonyms:
ethyl P-toluate
Molecular Weight (g/mol):
164.2
Molecular Formula:
C10H12O2
InChi Key:
NWPWRAWAUYIELB-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
Ethyl p-toluate, 98+%
CAS: 94-08-6 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.204 MDL Number: MFCD00009117 InChI Key: NWPWRAWAUYIELB-UHFFFAOYSA-N Synonym: ethyl p-toluate,ethyl p-methylbenzoate,benzoic acid, 4-methyl-, ethyl ester,4-methylbenzoic acid ethyl ester,unii-xuf0sq8l2j,p-toluic acid ethyl ester,xuf0sq8l2j,p-toluic acid, ethyl ester,4-methyl-benzoic acid ethyl ester,ethyl4-methylbenzoate PubChem CID: 66743 IUPAC Name: ethyl 4-methylbenzoate SMILES: CCOC(=O)C1=CC=C(C=C1)C