accessibility menu, dialog, popup

UserName

CAS RN 94-67-7

NOHOH(E)

CAS RN 94-67-7

IUPAC Name: (6Z)-6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
Synonyms: salicylaldoxime
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: GFCNBJDXQMZOOC-WAYWQWQTSA-N
SMILES: ON\C=C1\C=CC=CC1=O
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)