accessibility menu, dialog, popup

UserName

CAS RN 944-73-0

CH3NONCH3HNOHNO

CAS RN 944-73-0

IUPAC Name: 1,3-dimethyl-2,3,6,7,8,9-hexahydro-1H-purine-2,6,8-trione
Synonyms: 1,3-dimethyluric acid
Molecular Weight (g/mol): 196.17
Molecular Formula: C7H8N4O3
InChi Key: OTSBKHHWSQYEHK-UHFFFAOYSA-N
SMILES: CN1C2=C(NC(=O)N2)C(=O)N(C)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results