accessibility menu, dialog, popup

UserName

CAS RN 944450-83-3

CH3NNOHNH3C

CAS RN 944450-83-3

IUPAC Name: methyl({[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methyl})amine
Synonyms: methyl({[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 203.25
Molecular Formula: C11H13N3O
InChi Key: PBJZWGQLXKZEJT-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(C=C1)C1=NN=C(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results