CAS RN 947686-09-1
CAS RN 947686-09-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Duloxetine Phenyl Carbamate (>85%), TRC
CAS: 947686-09-1 Molecular Formula: C25H23NO3S Molecular Weight (g/mol): 417.52 Synonym: N-Methyl-N-[(3S)-3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]carbamic Acid Phenyl Ester,(S)-Duloxetine Impurity A; IUPAC Name: phenyl N-methyl-N-[(3S)-3-naphthalen-1-yloxy-3-thiophen-2-ylpropyl]carbamate SMILES: Oc1ccc(cc1)/C=C/C(=O)Nc2c(cccc2)CCC3N(C)CCCC3