accessibility menu, dialog, popup

UserName

CAS RN 94838-88-7

BOOOOHOH

CAS RN 94838-88-7

IUPAC Name: (6-formyl-2H-1,3-benzodioxol-5-yl)boronic acid
Synonyms: 6-formyl-2H-1,3-benzodioxol-5-ylboronic acid
Molecular Weight (g/mol): 193.95
Molecular Formula: C8H7BO5
InChi Key: AKEPUIJBVDGLMX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=C(OCO2)C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results