accessibility menu, dialog, popup

UserName

NHONH2OHH3C

CAS RN 951-55-3

IUPAC Name: 2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid
Synonyms: 5-methyltryptophan
Molecular Weight (g/mol): 218.26
Molecular Formula: C12H14N2O2
InChi Key: HUNCSWANZMJLPM-UHFFFAOYNA-N
SMILES: CC1=CC=C2NC=C(CC(N)C(O)=O)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results