accessibility menu, dialog, popup

UserName

CAS RN 959-22-8

ONOOO

CAS RN 959-22-8

IUPAC Name: 4-nitrophenyl benzoate
Synonyms: 4-nitrophenyl benzoate
Molecular Weight (g/mol): 243.22
Molecular Formula: C13H9NO4
InChi Key: GMKZBFFLCONHDE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(=O)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results