accessibility menu, dialog, popup

UserName

BrBr

CAS RN 959-88-6

IUPAC Name: 1-bromo-4-[4-(4-bromophenyl)buta-1,3-diyn-1-yl]benzene
Synonyms: 1-bromo-4-[4-(4-bromophenyl)buta-1,3-diyn-1-yl]benzene
Molecular Weight (g/mol): 360.05
Molecular Formula: C16H8Br2
InChi Key: IIERBCCHBFZMQZ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C#CC#CC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results