accessibility menu, dialog, popup

UserName

OO(S)NHOO

CAS RN 96402-49-2

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(naphthalen-1-yl)propanoate
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(naphthalen-1-yl)propanoate
Molecular Weight (g/mol): 436.49
Molecular Formula: C28H22NO4
InChi Key: ORWNVJDLEMVDLV-SANMLTNESA-M
SMILES: [O-]C(=O)[C@H](CC1=C2C=CC=CC2=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results