accessibility menu, dialog, popup

UserName

CH3HNNH2NCH3(E/Z)

CAS RN 97-39-2

IUPAC Name: N,N''-bis(2-methylphenyl)guanidine
Synonyms: ditolylguanidine
Molecular Weight (g/mol): 239.32
Molecular Formula: C15H17N3
InChi Key: OPNUROKCUBTKLF-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NC(N)=NC1=CC=CC=C1C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)