accessibility menu, dialog, popup

UserName

CAS RN 97108-50-4

H2NNHFF

CAS RN 97108-50-4

IUPAC Name: (2,5-difluorophenyl)hydrazine
Synonyms: (2,5-difluorophenyl)hydrazine
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H6F2N2
InChi Key: JNBRDOQHPXUXLY-UHFFFAOYSA-N
SMILES: NNC1=CC(F)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results