CAS RN 98-64-6
IUPAC Name:
4-chlorobenzene-1-sulfonamide
Synonyms:
4-chlorobenzenesulfonamide
Molecular Weight (g/mol):
191.63
Molecular Formula:
C6H6ClNO2S
InChi Key:
HHHDJHHNEURCNV-UHFFFAOYSA-N
SMILES:
NS(=O)(=O)C1=CC=C(Cl)C=C1
1
–
3
of
3
results
3
Alfa Aesar™ 4-Chlorobenzenesulfonamide, 98%
CAS: 98-64-6 Molecular Formula: C6H6ClNO2S Molecular Weight (g/mol): 191.63 MDL Number: MFCD00007936 InChI Key: HHHDJHHNEURCNV-UHFFFAOYSA-N Synonym: p-chlorobenzenesulfonamide,benzenesulfonamide, 4-chloro,p-chlorobenzenesulfamide,4-chlorophenylsulfonamide,p-chlorophenylsulfonamide,4-chlorobenzenesulphonamide,benzenesulfonamide, p-chloro,usaf ma-3,4-chlorobenzene-1-sulfonamide,4-chloro-benzenesulfonamide PubChem CID: 66824 IUPAC Name: 4-chlorobenzene-1-sulfonamide SMILES: NS(=O)(=O)C1=CC=C(Cl)C=C1