accessibility menu, dialog, popup

UserName

CAS RN 98045-03-5

CH3CH3H3COOHNOOHOOCH3

CAS RN 98045-03-5

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-4-methoxy-4-oxobutanoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-4-methoxy-4-oxobutanoic acid
Molecular Weight (g/mol): 247.25
Molecular Formula: C10H17NO6
InChi Key: IWFIVTBTZUCTQH-UHFFFAOYNA-N
SMILES: COC(=O)C(CC(O)=O)NC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)