CAS RN 98088-04-1
CAS RN 98088-04-1
IUPAC Name:
2-(pyrrolidin-1-yl)pyridine-4-carboxylic acid
Synonyms:
2-(pyrrolidin-1-yl)pyridine-4-carboxylic acid
Molecular Weight (g/mol):
192.22
Molecular Formula:
C10H12N2O2
InChi Key:
PQLHQGBNMJTAEH-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(=NC=C1)N1CCCC1
1
–
1
of
1
results
1
2-Pyrrolidin-1-ylisonicotinic acid, ≥90%, Thermo Scientific™
CAS: 98088-04-1 Molecular Formula: C10H12N2O2 Molecular Weight (g/mol): 192.22 MDL Number: MFCD04115212 InChI Key: PQLHQGBNMJTAEH-UHFFFAOYSA-N Synonym: 2-pyrrolidin-1-yl-isonicotinic acid,2-pyrrolidin-1-yl isonicotinic acid,2-pyrrolidin-1-yl pyridine-4-carboxylic acid,4-pyridinecarboxylic acid, 2-1-pyrrolidinyl,2-pyrrolidin-1-ylisonicotinic acid,2-1-pyrrolidinyl isonicotinic acid,2-1-pyrrolidinyl-4-pyridinecarboxylic acid,2-pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate,2-pyrrolidinylpyridine-4-carboxylic acid,hydrochloride PubChem CID: 6484236 IUPAC Name: 2-pyrrolidin-1-ylpyridine-4-carboxylic acid SMILES: OC(=O)C1=CC(=NC=C1)N1CCCC1