accessibility menu, dialog, popup

UserName

CAS RN 99-15-0

H3CCH3NHCH3OOHO

CAS RN 99-15-0

IUPAC Name: 2-acetamido-4-methylpentanoic acid
Synonyms: acetylleucine
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: WXNXCEHXYPACJF-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(C)=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)