accessibility menu, dialog, popup

UserName

CAS RN 99314-44-0

H3CSOOOOCH3

CAS RN 99314-44-0

IUPAC Name: (3-methyloxetan-3-yl)methyl 4-methylbenzene-1-sulfonate
Synonyms: (3-methyloxetan-3-yl)methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 256.32
Molecular Formula: C12H16O4S
InChi Key: PZOQQSOZRVZCMC-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC1(C)COC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results