accessibility menu, dialog, popup

UserName

CAS RN 994-89-8

CH3SnH3CCH3HC

CAS RN 994-89-8

IUPAC Name: tributyl(ethynyl)stannane
Synonyms: tributyl(ethynyl)stannane
Molecular Weight (g/mol): 315.09
Molecular Formula: C14H28Sn
InChi Key: YEMJHNYABQHWHL-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C#C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)