Summary of Key Physical Data for Solvents
Solvent Group
A classification for solvents in which they are placed in similar groups of selectivity.
Solvents of similar functionality are grouped into the same region of selectivity.
Group
1 Aliphatic ethers, tetramethylguanidine, hexamethyl phosphoric acid amides, trialkylamines
2 Aliphatic alcohols
3 Pyridine derivatives, tetrahydrofuran, amides (except formamide), glycol ethers, sulfoxides
4 Glycols, benzyl alcohol, acetic acid, formamide
5 Methylene chloride, ethylene chloride
6a Tricresyl phosphate, aliphatic ketones and esters, poly-ethers, dioxane
6b Sulfones, nitriles, propylene carbonate
7 Aromatic hydrocarbons, halo-substituted aromatic hydrocarbons, nitro compounds, aromatic ethers
8 Fluoroalkanols, m-cresol, water, chloroform
Polarity Index (p1)
Defined as a measure of the ability of the solvent
to interact with various test solutes. p1 increases
with solvent polarity.
References
• Classification of the Solvent Properties of Common Liquids.
L.R. Snyder.
Journal of Chromatography,
92 (1978) 223-234.
 Solvent   Solvent Group  Polarity Index (p1) Boiling Point (°C)   Viscosity 20°C (cP)  Refractive Index   Density at 20°C (g/mL)  
 Acetone   6a  5,156,20,321,3590,79
 Acetonitrile   6b  5,881,60,361,3440,781
 Benzene  72,780,10,651,5010,879
 Carbon tetrachloride   –  1,676,80,971,461,594
 1-Chlorobutane  5178,40,451,4020,886
 Chloroform  84,161,70,571,4441,489
 Cyclohexane   –  0,280,711,4270,778
 1,2 Dichloroethane  53,583,50,791,4451,253
 Dichloromethane  53,139,70,431,4241,324
 Diethyl ether  12,834,50,231,3520,713
 Dimethylformamide  36,41530,851,4290,949
 Dimethylsulfoxide  37,21892,241,4781,1
 1,4 Dioxane   6a  4,8101,11,541,4221,034
 Ethanol, absolute  24,3781,21,3610,789
 Ethyl acetate   6a  4,477,10,451,3720,9
 Heptane   –  0,1 94-97,5  0,411,3940,69
 Hexane   –  0,1 67-69,5  0,31 –  0,672
 Methanol  25,164,70,551,3280,791
 2-Methoxyethanol  35,5124,61,721,4020,965
 Methyl-tert-butyl ether  12,555,30,271,3690,741
 Octan-1-ol  23,4195,2 10,64 approx  1,4290,823
 Pentane   –   –  36,10,231,3570,626
 Petroleum ether   –  0,1 40-60, 60-80  0,3 –   0,64, 0,68  
 Propan-1-ol  2497,22,261,3850,804
 Propan-2-ol  23,982,32,31,3770,785
 Pyridine  35,3115,30,951,5090,982
 Tetrachloroethylene   –   –  121,20,931,5051,621
 Tetrahydrofuran  34660,551,4070,888
 Toluene  72,4110,60,591,4960,865
 1,2,4 Trichlorobenzene  7 –  214 –  1,5721,454
 2,2,4 Trimethylpentane –  0,199,20,511,3910,692
 Water  810,210011,3331