missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Pregnadien-17alpha,21-diol-3,11,20-trione-2,4,6alpha,9,12,12,21,21-d8, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Prednisone-d8 |
| Molecular Formula | C21 D8 H18 O5 |
| Quantity | 0.01 g |
| Synonym | Prednisone-d8, Prednisone-2,4,6α,9,12,12,21,21-D8, 17,21-Dihydroxypregna-1,4-diene-3,11,20-trione-2,4,6α,9,12,12,21,21-d8, 1,2-Dehydrocortisone-d8, 1,4-Pregnadien-17α,21-diol-3,11,20-trione-2,4,6α,9,12,12,21,21-d8 |
| InChI Formula | InChI=1 S/C21H26O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-15,18,22,26 H,3-4,6,8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1/i3D,5 D,9 D,10D2,11D2,18 D/t3-,14-,15-,18+,19-,20-,21- |
| SMILES | [2 H][C@H]1 C[C@H]2 [C@@H]3 CC[C@](O)(C(=O)C([2 H])([2 H])O)[C@@]3(C)C([2 H])([2 H])C(=O)[C@]2([2 H])[C@@]4(C)C=C([2 H])C(=O)C(=C14)[2 H] |
| IUPAC Name | (6 S,8 S,9 S,10 R,13 S,14 S,17 R)-2,4,6,9,12,12-hexadeuterio-17-(2,2-dideuterio-2-hydroxyacetyl)-17-hydroxy-10,13-dimethyl-6,7,8,14,15,16-hexahydrocyclopenta[a]phenanthrene-3,11-dione |
| Molecular Weight (g/mol) | 366.4774 |
| Formula Weight | 366.2282 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction