missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17alpha-Hydroxypregnenolone-21,21,21-d3, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 17 A-Hydroxypregnenolone-d3 |
| CAS | 105078-92-0 |
| Molecular Formula | C21 2H3 H29 O3 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C21H32O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h4,15-18,23-24 H,5-12H2,1-3H3/t15-,16+,17-,18-,19-,20-,21-/m0/s1/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)[C@@]1(O)CC[C@H]2 [C@@H]3 CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | 2,2,2-trideuterio-1-[(3 S,8 R,9 S,10 R,13 S,14 S,17 R)-3,17-dihydroxy-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl]ethanone |
| Molecular Weight (g/mol) | 335.5 |
| Formula Weight | 335.254 g/mol |
| Percent Purity | 97 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction