missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethylpropionyl-L-carnitine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Pivaloyl-L-carnitine-D3 (N-methyl-D3) Chloride |
| Molecular Formula | C12 D3 H21 N O4 . Cl |
| Quantity | 0.01 g |
| Synonym | Pivaloyl-L-carnitine-d3 chloride, Pivaloyl-L-carnitine-D3 (N-methyl-D3) Chloride, (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-(2,2-dimethyl-1-oxopropoxy)-1-propanaminium chloride, [(2 R)-3-Carboxy-2-(2,2-dimethylpropanoyloxy)propyl]dimethyl(methyl-d3)ammonium chloride, 2,2-Dimethylpropionyl-L-carnitine-d3 HCl (N-methyl-d3) |
| InChI Formula | InChI=1 S/C12H23NO4.ClH/c1-12(2,3)11(16)17-9(7-10(14)15)8-13(4,5)6;/h9H,7-8H2,1-6H3;1 H/t9-;/m1./s1/i4D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)C(C)(C)C |
| IUPAC Name | [(2 R)-3-carboxy-2-(2,2-dimethylpropanoyloxy)propyl]-dimethyl-(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 284.7948 |
| Formula Weight | 284.1582 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion