Learn More
2'deoxycytidine-5,6-d2, CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 2'-Deoxycytidine-5,6-D2 |
| CAS | 1186526-91-9 |
| Molecular Formula | C9 D2 H11 N3 O4 |
| Quantity | 0.01 g |
| Synonym | Cytidine-5,6-d2, 2'-deoxy-, 2'-Deoxycytidine-5,6-D2, 5,6-[2 H] 2'-Deoxycytidine |
| InChI Formula | InChI=1 S/C9H13N3O4/c10-7-1-2-12(9(15)11-7)8-3-5(14)6(4-13)16-8/h1-2,5-6,8,13-14 H,3-4H2,(H2,10,11,15)/t5-,6+,8+/m0/s1/i1D,2 D |
| SMILES | [2 H]C1=C([2 H])C(=NC(=O)N1[C@H]2 C[C@H](O)[C@@H](CO)O2)N |
| IUPAC Name | 4-amino-5,6-dideuterio-1-[(2 R,4 S,5 R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
| Molecular Weight (g/mol) | 229.2295 |
| Formula Weight | 229.1032 g/mol |
| Show More |