Learn More
3-(Methyl-d3-thio)propionaldehyde, CDN
Specifications
Specifications
| Chemical Name or Material | 3-Methyl-d3-thiopropanal |
| CAS | 136430-27-8 |
| Molecular Formula | C4 2H3 H5 S |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C4H8S/c1-2-3-4-5/h4H,2-3H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCC=S |
| IUPAC Name | 4,4,4-trideuteriobutanethial |
| Molecular Weight (g/mol) | 91.19 |
| Formula Weight | 91.0535 g/mol |
| Percent Purity | 98 atom % D, min 96% Chemical Purity |