missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Pregnadien-6-methyl-16-methylene-17a-ol-3,20-dione Acetate-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Melengestrol acetate-d3 |
| Molecular Formula | C25 D3 H29 O4 |
| Quantity | 0.01 g |
| Synonym | 17-(Acetyloxy-d3)-6-methyl-16-methylenepregna-4,6-diene-3,20-dione, 17-Hydroxy-6-methyl-16-methylenepregna-4,6-diene-3,20-dione Acetate-d3, 17-(Acetoxy-d3)-6-methyl-16-methylenepregna-4,6-diene-3,20-dione, 6-Dehydro-16-methylene-17α-hydroxy-6α-methylprogesterone Acetate-d3, Heifermax 500-d3, 4,6-Pregnadien-6-methyl-16-methylene-17 A-ol-3,20-dione Acetate-d3, 4,6-Pregnadien-6-methyl-16-methylene-17 A-ol-3,20-dione Acetate-d3, 4,6-Pregnadien-6-methyl-16-methylene-17 A-ol-3,20-dione Acetate-d3 |
| InChI Formula | InChI=1 S/C25H32O4/c1-14-11-19-20(23(5)9-7-18(28)13-21(14)23)8-10-24(6)22(19)12-15(2)25(24,16(3)26)29-17(4)27/h11,13,19-20,22 H,2,7-10,12H2,1,3-6H3/t19-,20+,22+,23-,24+,25+/m1/s1/i4D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)O[C@]1(C(=O)C)C(=C)C[C@H]2 [C@@H]3 C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 R)-17-acetyl-6,10,13-trimethyl-16-methylidene-3-oxo-1,2,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-17-yl] 2,2,2-trideuterioacetate |
| Molecular Weight (g/mol) | 399.54 |
| Formula Weight | 399.2489 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction