missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouracil-15N2, CDN

Product Code. 30316661
Click to view available options
Quantity:
0.005 g
0.01 g
This item is not returnable. View return policy

Product Code. 30316661

Brand: CDN CDNN21400.005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Fluorouracil-15N2
CAS 68941-95-7
Molecular Formula C4H3F(15 N)2O2
Quantity 0.005 g
Synonym 5-Fluoropyrimidine-2,4(1 H,3 H)-dione-1,3-15N2
InChI Formula InChI=1 S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)/i6+1,7+1
SMILES O=C1C(F)=C[15 NH]C(=O)[15 NH]1
IUPAC Name 5-fluoropyrimidine-2,4(1 H,3 H)-dione-1,3-15N2
Molecular Weight (g/mol) 132.06
Formula Weight 132.01 g/mol
Percent Purity 98 atom % 15 N, min 98% Chemical Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.