Learn More
5-Hydroxyindole-3-acetic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxyindole-3-acetic acid 2,2-d2 |
| CAS | 56209-31-5 |
| Molecular Formula | C10 2H2 H7 N O3 |
| Quantity | 0.01 g |
| Synonym | 1 H-Indole-3-acetic-α,α-d2 acid, 5-hydroxy- (9 CI), 5-Hydroxy-1 H-indole-3-acetic-α,α-d2 acid, 5-Hydroxyindole-3-acetic-2,2-d2 acid, 5-Hydroxyindole-3-acetic acid-d2, 5-Hydroxyindole-3-acetic-2,2-D2 acid, 5-HIAA-d2 |
| InChI Formula | InChI=1 S/C10H9NO3/c12-7-1-2-9-8(4-7)6(5-11-9)3-10(13)14/h1-2,4-5,11-12 H,3H2,(H,13,14)/i3D2 |
| SMILES | [2 H]C([2 H])(C(=O)O)c1c[nH]c2ccc(O)cc12 |
| IUPAC Name | 2,2-dideuterio-2-(5-hydroxy-1 H-indol-3-yl)acetic acid |
| Molecular Weight (g/mol) | 193.2 |
| Formula Weight | 193.0708 g/mol |
| Show More |