Learn More
(±)-Carvedilol-d4 (ethyl-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Carvedilol-D4 (2-aminoethoxy-1,1,2,2-D4) |
| CAS | 1133705-56-2 |
| Molecular Formula | C24 D4 H22 N2 O4 |
| Quantity | 0.005 g |
| Synonym | 2-Propanol, 1-(9 H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl-1,1,2,2-d4]amino]- (ACI), 1-(9 H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl-1,1,2,2-d4]amino]-2-propanol (ACI), 1-(9 H-Carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)-1,1,2,2-tetradeuterioethylamino]propan-2-ol, Carvedilol-d4, Carvedilol-D4 (2-aminoethoxy-1,1,2,2-D4), (±)-Carvedilol-d4 (ethyl-d4) |
| InChI Formula | InChI=1 S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27 H,13-16H2,1H3/i13D2,14D2 |
| SMILES | [2 H]C([2 H])(NCC(O)COc1cccc2[nH]c3ccccc3c12)C([2 H])([2 H])Oc4ccccc4OC |
| IUPAC Name | 1-(9 H-carbazol-4-yloxy)-3-[[1,1,2,2-tetradeuterio-2-(2-methoxyphenoxy)ethyl]amino]propan-2-ol |
| Molecular Weight (g/mol) | 410.4989 |
| Formula Weight | 410.2144 g/mol |
| Show More |