Learn More
Chlorambucil-d8 [bis(2-chloroethyl-d4)], CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Chlorambucil-D8 (bis(2-chloroethyl-D4)) |
| CAS | 2748247-26-7 |
| Molecular Formula | C14 D8 H11 Cl2 N O2 |
| Quantity | 0.005 g |
| Synonym | Chlorambucil-d8 [bis(2-chloroethyl-d4)], Chlorambucil-D8 (bis(2-chloroethyl-D4)), Chlorambucil-d8, 4-[Bis(2-chloroethyl-1,1,2,2-d4)amino]benzenebutanoic acid |
| InChI Formula | InChI=1 S/C14H19Cl2NO2/c15-8-10-17(11-9-16)13-6-4-12(5-7-13)2-1-3-14(18)19/h4-7 H,1-3,8-11H2,(H,18,19)/i8D2,9D2,10D2,11D2 |
| SMILES | [2 H]C([2 H])(Cl)C([2 H])([2 H])N(c1ccc(CCCC(=O)O)cc1)C([2 H])([2 H])C([2 H])([2 H])Cl |
| IUPAC Name | 4-[4-[bis(2-chloro-1,1,2,2-tetradeuterioethyl)amino]phenyl]butanoic acid |
| Molecular Weight (g/mol) | 312.2615 |
| Formula Weight | 311.1295 g/mol |
| Show More |