missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlormequat-1,1,2,2-d4 Chloride, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Chlormequat-chloride 1,1,2,2-D4 |
| Molecular Formula | C5 D4 H9 Cl N . Cl |
| Quantity | 0.01 g |
| Synonym | Chlormequat-chloride 1,1,2,2-D4, Chlormequat Chloride D4 (chloroethyl D4), Chlormequat chloride-d4, Chlormequat Chloride D4 (1,1,2,2-D4) |
| InChI Formula | InChI=1 S/C5H13ClN.ClH/c1-7(2,3)5-4-6;/h4-5H2,1-3H3;1 H/q+1;/p-1/i4D2,5D2; |
| SMILES | [Cl-].[2 H]C([2 H])(Cl)C([2 H])([2 H])[N+](C)(C)C |
| IUPAC Name | (2-chloro-1,1,2,2-tetradeuterioethyl)-trimethylazanium;chloride |
| Molecular Weight (g/mol) | 162.094 |
| Formula Weight | 161.068 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction