missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ciprofloxacin-d8 HCl H2O (piperazine-d8), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Ciprofloxacin-D8 (piperazine-D8) Hydrochloride Monohydrate |
| Molecular Formula | C17 D8 H10 F N3 O3 . Cl H . H2 O |
| Quantity | 0.005 g |
| Synonym | 1-Cyclopropyl-6-fluoro-7-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid hydrochloride monohydrate, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl-2,2,3,3,5,5,6,6-d8)-3-quinolinecarboxylic acid hydrochloride hydrate (1:1:1), Ciprofloxacin-d8 hydrochloride monohydrate, Ciprofloxacin-D8 (piperazine-D8) hydrochloride monohydrate, Ciprofloxacin-d8 HCl H2O (piperazine-d8) |
| InChI Formula | InChI=1 S/C17H18FN3O3.ClH.H2O/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24;;/h7-10,19 H,1-6H2,(H,23,24);1 H;1H2/i3D2,4D2,5D2,6D2;; |
| SMILES | O.Cl.[2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])N(c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)C4CC4)C1([2 H])[2 H] |
| IUPAC Name | 1-cyclopropyl-6-fluoro-7-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrate;hydrochloride |
| Molecular Weight (g/mol) | 393.867 |
| Formula Weight | 393.1707 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion