Learn More
Cortisol-9,11,12,12-d4, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Cortisol-D4 (9,11,12,12-D4) |
| CAS | 73565-87-4 |
| Molecular Formula | C21 2H4 H26 O5 |
| Quantity | 0.005 g |
| Synonym | Hydrocortisone-9α,11α,12α,12β-D4 |
| InChI Formula | InChI=1 S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26 H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1/i10D2,16 D,18 D |
| SMILES | [2 H]C1([2 H])[C@]([2 H])(O)[C@@]2([2 H])[C@@H](CCC3=CC(=O)CC[C@]23 C)[C@@H]4 CC[C@](O)(C(=O)CO)[C@@]14 C |
| IUPAC Name | (8 S,9 S,10 R,11 S,13 S,14 S,17 R)-9,11,12,12-tetradeuterio-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,14,15,16-octahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 366.48 |
| Formula Weight | 366.2344 g/mol |
| Show More |