missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dabigatran Etexilate-d13 (hexyloxy-d13), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Dabigatran Etexilate-D13 (hexyloxy-D13) |
| CAS | 2749293-92-1 |
| Molecular Formula | C34 D13 H28 N7 O5 |
| Quantity | 0.01 g |
| Synonym | N-[[2-[[[4-[Amino[[(hexyloxy-d13)carbonyl]imino]methyl]phenyl]amino]methyl]-1-methyl-1 H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-β-alanine ethyl ester, Dabigatran Etexilate-d13, Dabigatran Etexilate-D13 (hexyloxy-D13) |
| InChI Formula | InChI=1 S/C34H41N7O5/c1-4-6-7-10-21-46-34(44)39-32(35)24-12-15-26(16-13-24)37-23-30-38-27-22-25(14-17-28(27)40(30)3)33(43)41(20-18-31(42)45-5-2)29-11-8-9-19-36-29/h8-9,11-17,19,22,37 H,4-7,10,18,20-21,23H2,1-3H3,(H2,35,39,44)/i1D3,4D2,6D2,7D2,10D2,21D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OC(=O)NC(=N)c1ccc(NCc2nc3cc(ccc3n2C)C(=O)N(CCC(=O)OCC)c4ccccn4)cc1 |
| IUPAC Name | ethyl 3-[[1-methyl-2-[[4-[N-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecadeuteriohexoxycarbonyl)carbamimidoyl]anilino]methyl]benzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate |
| Molecular Weight (g/mol) | 640.8133 |
| Formula Weight | 640.3985 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction