missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Dapoxetine-d6 HCl (N,N-dimethyl-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (+/-)-Dapoxetine-D6 (N,N-dimethyl-D6) Hydrochloride |
| Molecular Formula | C21 D6 H17 N O . Cl H |
| Quantity | 0.01 g |
| Synonym | N,N-Di(methyl-d3)-α-[2-(1-naphthalenyloxy)ethyl]benzenemethanamine hydrochloride (1:1), 3-Naphthalen-1-yloxy-1-phenyl-N,N-bis(trideuteriomethyl)propan-1-amine hydrochloride, (±)-Dapoxetine-d6 hydrochloride, (±)-Dapoxetine-D6 (N,N-dimethyl-D6) hydrochloride, N,N-Di(methyl-d3)-3-(1-naphthoxy)-1-phenyl-1-propylamine monohydrochloride |
| InChI Formula | InChI=1 S/C21H23NO.ClH/c1-22(2)20(18-10-4-3-5-11-18)15-16-23-21-14-8-12-17-9-6-7-13-19(17)21;/h3-14,20 H,15-16H2,1-2H3;1 H/i1D3,2D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N(C(CCOc1cccc2ccccc12)c3ccccc3)C([2 H])([2 H])[2 H] |
| IUPAC Name | 3-naphthalen-1-yloxy-1-phenyl-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 347.9113 |
| Formula Weight | 347.1923 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction