Learn More
(±)-Fenfluramine-d5 HCl (N-ethyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | rac Fenfluramine-d5 Hydrochloride |
| CAS | 1216927-29-5 |
| Molecular Formula | C12 D5 H11 F3 N . Cl H |
| Quantity | 0.005 g |
| Synonym | Fenfluramine Hydrochloride D5 (Ethyl D5), N-(Ethyl-d5)-α-methyl-3-(trifluoromethyl)benzeneethanamine hydrochloride, 3-(Trifluoromethyl)-N-(ethyl-d5)-α-methylphenethylamine hydrochloride, N-Pentadeuterioethyl-α-methyl-3-(trifluoromethyl)phenethylamine hydrochloride, N-(1,1,2,2,2-Pentadeuterioethyl)-3-[3-(trifluoromethyl)phenyl]propan-2-amine hydrochloride, Fenfluramine-d5 hydrochloride, Fenfluramine-D5 (N-ethyl-D5) hydrochloride |
| InChI Formula | InChI=1 S/C12H16F3N.ClH/c1-3-16-9(2)7-10-5-4-6-11(8-10)12(13,14)15;/h4-6,8-9,16 H,3,7H2,1-2H3;1 H/i1D3,3D2; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])NC(C)Cc1cccc(c1)C(F)(F)F |
| IUPAC Name | N-(1,1,2,2,2-pentadeuterioethyl)-1-[3-(trifluoromethyl)phenyl]propan-2-amine;hydrochloride |
| Molecular Weight (g/mol) | 272.749 |
| Formula Weight | 272.132 g/mol |
| Show More |