Learn More
Lidocaine-d10 (N,N-diethyl-d10), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Lidocaine N,N-diethyl-d10 |
| CAS | 851528-09-1 |
| Molecular Formula | C14 2H10 H12 N2 O |
| Quantity | 0.01 g |
| Synonym | Lidocaine-D10 (N,N-di(ethyl-D5)), Acetamide, 2-[di(ethyl-d5)amino]-N-(2,6-dimethylphenyl)- (9 CI), 2-(Bis(1,1,2,2,2-pentadeuterioethyl)amino)-N-(2,6-dimethylphenyl)acetamide, Lidocaine-d10, α-(Diethylamino-d10)-2,6-acetoxylidide, 2-[Di(ethyl-d5)amino]-2',6'-dimethylacetanilide, 2-[Di(ethyl-d5)amino]-2',6'-dimethylacetanilide, 2-[Di(ethyl-d5)amino]-2',6'-dimethylacetanilide |
| InChI Formula | InChI=1 S/C14H22N2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4/h7-9 H,5-6,10H2,1-4H3,(H,15,17)/i1D3,2D3,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])N(CC(=O)Nc1c(C)cccc1C)C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-[bis(1,1,2,2,2-pentadeuterioethyl)amino]-N-(2,6-dimethylphenyl)acetamide |
| Molecular Weight (g/mol) | 244.4 |
| Formula Weight | 244.236 g/mol |
| Show More |