Learn More
Malathion-d7 (dimethyl-d6; 3-d1), CDN
Product Code. 30310330
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30310330
Brand: CDN CDND51780.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Malathion D7 (dimethyl D6,3-D1)
CAS 352438-94-9
Molecular Formula C10 2H7 H12 O6 P S2
Quantity 0.01 g
Synonym Butanedioic-d acid, 3-[[di(methoxy-d3)phosphinothioyl]thio]-, diethyl ester (9 CI), Malathion D7 (dimethyl D6,3-D1)
InChI Formula InChI=1 S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3/i3D3,4D3,7 D
SMILES [2 H]C(C(SP(=S)(OC([2 H])([2 H])[2 H])OC([2 H])([2 H])[2 H])C(=O)OCC)C(=O)OCC
IUPAC Name diethyl 2-[bis(trideuteriomethoxy)phosphinothioylsulfanyl]-3-deuteriobutanedioate
Molecular Weight (g/mol) 337.4
Formula Weight 337.08 g/mol
Percent Purity 99 atom % D, min 95% Chemical Purity