missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Mepivacaine-d3 (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Mepivacaine-d3 |
| CAS | 1346597-90-7 |
| Molecular Formula | C15 D3 H19 N2 O |
| Quantity | 0.01 g |
| Synonym | N-(2,6-Dimethylphenyl)-1-(trideuteriomethyl)piperidine-2-carboxamide, N-(2,6-Dimethylphenyl)-1-(methyl-d3)-2-piperidinecarboxamide, 1-(Methyl-d3)-2',6'-pipecoloxylidide, (±)-Mepivacaine-d3, Mepivacaine-d3, (±)-Mepivacaine-d3 (N-methyl-d3), (±)-Mepivacaine-d3 (N-methyl-d3) |
| InChI Formula | InChI=1 S/C15H22N2O/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3/h6-8,13 H,4-5,9-10H2,1-3H3,(H,16,18)/i3D3 |
| SMILES | [2 H]C([2 H])([2 H])N1CCCCC1C(=O)Nc2c(C)cccc2C |
| IUPAC Name | N-(2,6-dimethylphenyl)-1-(trideuteriomethyl)piperidine-2-carboxamide |
| Molecular Weight (g/mol) | 249.367 |
| Formula Weight | 249.192 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction