Learn More
N-(3-Methylbutyryl-d9)glycine, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | N-(3-Methylbutanoyl)glycine 3-Methylbutanoyl-d9 |
| CAS | 1330037-21-2 |
| Molecular Formula | C7 2H9 H4 N O3 |
| Quantity | 0.01 g |
| Synonym | Glycine, N-[3-(methyl-d3)-1-oxobutyl-2,2,3,4,4,4-d6]-, N-(3-(Methyl-d3)-1-oxobutyl-2,2,3,4,4,4-d6)glycine, Isovalerylglycine-d9, N-Isovaleroylglycine-d9, N-Isovalerylglycined-d9, N-(3-Methylbutanoyl)glycine-D9 (3-Methylbutanoyl-D9) |
| InChI Formula | InChI=1 S/C7H13NO3/c1-5(2)3-6(9)8-4-7(10)11/h5H,3-4H2,1-2H3,(H,8,9)(H,10,11)/i1D3,2D3,3D2,5 D |
| SMILES | C(NCC(O)=O)(C(C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])[2 H])([2 H])[2 H])=O |
| IUPAC Name | 2-[[2,2,3,4,4,4-hexadeuterio-3-(trideuteriomethyl)butanoyl]amino]acetic acid |
| Molecular Weight (g/mol) | 168.24 |
| Formula Weight | 168.146 g/mol |
| Show More |