Learn More
Nifedipine-d4 (2-nitrophenyl-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Nifedipine-D4 (2-nitrophenyl-D4) |
| CAS | 1219798-99-8 |
| Molecular Formula | C17 D4 H14 N2 O6 |
| Quantity | 0.01 g |
| Synonym | 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(6-nitrophenyl-2,3,4,5-d4)-, 3,5-dimethyl ester (ACI), 3,5-Dimethyl 1,4-dihydro-2,6-dimethyl-4-(6-nitrophenyl-2,3,4,5-d4)-3,5-pyridinedicarboxylate (ACI), 1,4-Dihydro-2,6-dimethyl-4-(6-nitrophenyl-2,3,4,5-d4)-3,5-pyridinedicarboxylic acid 3,5-dimethyl ester, Nifedipine-d4 |
| InChI Formula | InChI=1 S/C17H18N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8,15,18 H,1-4H3/i5D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c(C2C(=C(C)NC(=C2C(=O)OC)C)C(=O)OC)c1[2 H])[N+](=O)[O-] |
| IUPAC Name | dimethyl 2,6-dimethyl-4-(2,3,4,5-tetradeuterio-6-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| Molecular Weight (g/mol) | 350.3593 |
| Formula Weight | 350.1416 g/mol |
| Show More |