missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ractopamine-d6 HCl (1-methyl-d3-propyl-1,2,2-d3) (mixture of diastereomers), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Ractopamine-d6 hydrochloride |
| CAS | 1276197-17-1 |
| Molecular Formula | C18 2H6 H17 N O3 . Cl H |
| Quantity | 0.01 g |
| Synonym | Ractopamine D6 Hydrochloride, 4-[2-[[1,1,1,2,3,3-Hexadeuterio-4-(4-hydroxyphenyl)butan-2-yl]amino]-1-hydroxyethyl]phenol hydrochloride |
| InChI Formula | InChI=1 S/C18H23NO3.ClH/c1-13(2-3-14-4-8-16(20)9-5-14)19-12-18(22)15-6-10-17(21)11-7-15;/h4-11,13,18-22 H,2-3,12H2,1H3;1 H/i1D3,2D2,13 D; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])(NCC(O)c1ccc(O)cc1)C([2 H])([2 H])Cc2ccc(O)cc2 |
| IUPAC Name | 4-[2,2,3,4,4,4-hexadeuterio-3-[[2-hydroxy-2-(4-hydroxyphenyl)ethyl]amino]butyl]phenol;hydrochloride |
| Molecular Weight (g/mol) | 343.88 |
| Formula Weight | 343.1821 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction