missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-propyl-d28-ammonium Bromide, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Tetrapropylammonium-D28 Bromide |
| CAS | 284474-84-6 |
| Molecular Formula | C12 D27 H N . Br |
| Quantity | 0.01 g |
| Synonym | 1-Propan-1,1,2,2,3,3,3-d7-aminium, N,N,N-tri(propyl-d7)-, bromide (9 CI), Tetrapropylammonium-D28 bromide, N,N,N-Tri(propyl-d7)-1-propan-1,1,2,2,3,3,3-d7-aminium bromide, Tetrapropylammonium-d28 bromide, Tetra-n-propylammonium-d28 bromide |
| InChI Formula | InChI=1 S/C12H28N.BrH/c1-5-9-13(10-6-2,11-7-3)12-8-4;/h5-12H2,1-4H3;1 H/q+1;/p-1/i1D2,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2; |
| SMILES | [Br-].[2 H]C([2 H])C([2 H])([2 H])C([2 H])([2 H])[N+](C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])(C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | tris(1,1,2,2,3,3,3-heptadeuteriopropyl)-(1,1,2,2,3,3-hexadeuteriopropyl)azanium;bromide |
| Molecular Weight (g/mol) | 293.428 |
| Formula Weight | 292.31 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction