missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Theobromine-d3 (7-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Theobromine-d3 (7-methyl-d3) |
| CAS | 65566-69-0 |
| Molecular Formula | C7 D3 H5 N4 O2 |
| Quantity | 0.005 g |
| Synonym | Theobromine D3 (7-Methyl D3), 3,7-Dihydro-3-methyl-7-(methyl-d3)-1 H-purine-2,6-dione (ACI), 1 H-Purine-2,6-dione, 3,7-dihydro-3-methyl-7-(methyl-d3)- (9 CI, ACI) |
| InChI Formula | InChI=1 S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)9-7(13)11(5)2/h3H,1-2H3,(H,9,12,13)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])n1cnc2N(C)C(=O)NC(=O)c12 |
| IUPAC Name | 3-methyl-7-(trideuteriomethyl)purine-2,6-dione |
| Molecular Weight (g/mol) | 183.183 |
| Formula Weight | 183.084 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction