missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-Muconic-d4 Acid, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | trans,trans-Muconic Acid D4 |
| CAS | 1955496-83-9 |
| Molecular Formula | C6 D4 H2 O4 |
| Quantity | 0.005 g |
| Synonym | (2 E,4 E)-2,4-Hexadienedioic-2,3,4,5-d4 acid, (E,E)-Muconic acid-d4, trans,trans-1,2,3,4-Tetradeuterio-1,3-butadiene-1,4-dicarboxylic acid, trans,trans-Muconic-d4 Acid |
| InChI Formula | InChI=1 S/C6H6O4/c7-5(8)3-1-2-4-6(9)10/h1-4 H,(H,7,8)(H,9,10)/b3-1+,4-2+/i1D,2 D,3 D,4 D |
| SMILES | [2 H]\C(=C(\[2 H])/C(=O)O)\C(=C(/[2 H])\C(=O)O)\[2 H] |
| IUPAC Name | (2 E,4 E)-2,3,4,5-tetradeuteriohexa-2,4-dienedioic acid |
| Molecular Weight (g/mol) | 146.1341 |
| Formula Weight | 146.0517 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction