Learn More
1-Bromo-3-chloropropane-d6, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-3-chloropropane-D6 |
| CAS | 1173018-46-6 |
| Molecular Formula | C3 D6 Br Cl |
| Quantity | 0.05 g |
| Synonym | Propane-1,1,2,2,3,3-d6, 1-bromo-3-chloro- (ACI), 1-Bromo-3-chloropropane-1,1,2,2,3,3-d6 (ACI), 1-Bromo-3-chloropropane-d6, 1-Bromo-3-chloropropane-D6 |
| InChI Formula | InChI=1 S/C3H6BrCl/c4-2-1-3-5/h1-3H2/i1D2,2D2,3D2 |
| SMILES | [2 H]C([2 H])(Cl)C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1-bromo-3-chloro-1,1,2,2,3,3-hexadeuteriopropane |
| Molecular Weight (g/mol) | 163.4737 |
| Formula Weight | 161.9718 g/mol |
| Show More |