missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromo-4-cyanophenyl Octanoate-d15, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Bromoxynil Octanoate-D15 |
| Molecular Formula | C15 D15 H2 Br2 N O2 |
| Quantity | 0.05 g |
| Synonym | 2,6-Dibromo-4-cyanophenyl octanoate-d15, 3,5-Dibromo-4-hydroxybenzonitrile octanoate-d15 ester, 3,5-Dibromo-4-(octanoyloxy-d15)benzonitrile, 4-Cyano-2,6-dibromophenyl octanoate-d15, Bromoxynil Octanoate-D15, Bromoxynil octanoic-d15 acid ester |
| InChI Formula | InChI=1 S/C15H17Br2NO2/c1-2-3-4-5-6-7-14(19)20-15-12(16)8-11(10-18)9-13(15)17/h8-9 H,2-7H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)Oc1c(Br)cc(cc1Br)C#N |
| IUPAC Name | (2,6-dibromo-4-cyanophenyl) 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecadeuteriooctanoate |
| Molecular Weight (g/mol) | 418.2014 |
| Formula Weight | 416.0568 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction