missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-1,5-pentanedioic-2,2,3,4,4-d5 Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 3-Hydroxypentanedioic Acid D5 |
| CAS | 1219805-72-7 |
| Molecular Formula | C5H3D5O5 |
| Quantity | 0.05 g |
| Synonym | 3-Hydroxy-pentanedioic Acid-d5, 2,4-Dideoxypentaric Acid-d5, β-Hydroxyglutaric Acid-d5; |
| InChI Formula | InChI=1 S/C5H8O5/c6-3(1-4(7)8)2-5(9)10/h3,6 H,1-2H2,(H,7,8)(H,9,10)/i1D2,2D2,3 D |
| SMILES | OC(C([2 H])([2 H])C([2 H])(O)C([2 H])([2 H])C(O)=O)=O |
| IUPAC Name | 3-hydroxypentanedioic-2,2,3,4,4-d5 acid |
| Molecular Weight (g/mol) | 153.14 |
| Formula Weight | 153.07 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction