missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Chloro-2-methylphenoxy-d3)butyric Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 4-(4-Chloro-2-methylphenoxy-d3)butyric Acid |
| CAS | 1219803-42-5 |
| Molecular Formula | C11 D3 H10 Cl O3 |
| Quantity | 0.05 g |
| Synonym | 4-(4-Chloro-2-methylphenoxy-d3)butyric Acid, MCPB D3 (ring D3), 4-(4-Chloro-2-methylphenoxy)butanoic Acid (ring D3), 4-(4-Chloro-2-methylphenoxy-3,5,6-d3)butanoic acid |
| InChI Formula | InChI=1 S/C11H13ClO3/c1-8-7-9(12)4-5-10(8)15-6-2-3-11(13)14/h4-5,7 H,2-3,6H2,1H3,(H,13,14)/i4D,5 D,7 D |
| SMILES | [2 H]c1c([2 H])c(OCCCC(=O)O)c(C)c([2 H])c1Cl |
| IUPAC Name | 4-(4-chloro-2,3,5-trideuterio-6-methylphenoxy)butanoic acid |
| Molecular Weight (g/mol) | 231.6906 |
| Formula Weight | 231.0742 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction