Learn More
Caffeine-d9 (1,3,7-trimethyl-d9), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Caffeine-d9 |
| CAS | 72238-85-8 |
| Molecular Formula | C8 2H9 H N4 O2 |
| Quantity | 0.05 g |
| Synonym | 1 H-Purine-2,6-dione, 3,7-dihydro-1,3,7-tri(methyl-d3)-, 3,7-Dihydro-1,3,7-tri(methyl-d3)-1 H-purine-2,6-dione, Caffeine-d9 |
| InChI Formula | InChI=1 S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i1D3,2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])N1C(=O)N(c2ncn(c2C1=O)C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,3,7-tris(trideuteriomethyl)purine-2,6-dione |
| Molecular Weight (g/mol) | 203.25 |
| Formula Weight | 203.1369 g/mol |
| Show More |